
Methyl 5-bromo-1H-indazole-7-carboxylate
Catalog # APOSOR61032-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.068 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:898747-24-5
Formula:C₉H₇BrN₂O₂
MDL Number:MFCD08458991