Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-5-methyl-1H-indazole, N1-BOC protected
4-Bromo-5-methyl-1H-indazole, N1-BOC protected
4-Bromo-5-methyl-1H-indazole, N1-BOC protected
APOSOR61030-250MG
:
Apollo Scientific
:
4-Bromo-5-methyl-1H-indazole, N1-BOC protected
APOSOR61030-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
311.174 g/mol
:
GHS07
:
Refrigerator
:
926922-41-0
:
C₁₃H₁₅BrN₂O₂
:
MFCD16628190