Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-2-hydrazino-3-methoxypyrazine
6-Bromo-2-hydrazino-3-methoxypyrazine
6-Bromo-2-hydrazino-3-methoxypyrazine
6-Bromo-2-hydrazino-3-methoxypyrazine
Supplier:
Apollo Scientific
Certificates
Expand image
6-Bromo-2-hydrazino-3-methoxypyrazine
Supplier:
Apollo Scientific
About this item
CAS Number:
1334135-54-4
:
Formula:
C₅H₇BrN₄O
MDL Number:
MFCD22375534
:
219.039 g/mol
:
GHS07
Storage Temperature:
Refrigerator