0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
4-Hydroxyphenylacetaldehyde
4-Hydroxyphenylacetaldehyde
4-Hydroxyphenylacetaldehyde
Irritant
4-Hydroxyphenylacetaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Hydroxyphenylacetaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
7339-87-9
:
Formula:
C₈H₈O₂
MDL Number:
MFCD03411043
:
136.148 g/mol
:
GHS07
Storage Temperature:
Freezer