tert-Butyl 2-azaspiro[3.3]hept-6-ylcarbamate hemioxalate
Catalog # APOSOR55148-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:514.612 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1523572-07-7
Formula:2C₁₁H₂₀N₂O₂·C₂H₂O₄
MDL Number:MFCD22421679