Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-3-methylaniline
2-Chloro-3-methylaniline 97%
2-Chloro-3-methylaniline 97%
2-Chloro-3-methylaniline 97%
APOSOR55072-5G
:
Apollo Scientific
:
2-Chloro-3-methylaniline 97%
APOSOR55072-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
141.598 g/mol
:
GHS07
:
Ambient
:
29027-17-6
:
C₇H₈ClN
:
MFCD11113332