Order Entry
Germany
ContactUsLinkComponent
 
tert-Butyl 3-(4-aminophenyl)piperidine-1-carboxylate 95%
Catalog # APOSOR54550-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 3-(4-aminophenyl)piperidine-1-carboxylate 95%
Catalog # APOSOR54550-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :276.374 g/mol
Storage Temperature:Ambient
CAS Number:875798-79-1
Formula:C₁₆H₂₄N₂O₂
MDL Number:MFCD05861561