Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxypyridin-4-ol
2-Methoxypyridin-4-ol 95+%
2-Methoxypyridin-4-ol 95+%
2-Methoxypyridin-4-ol 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Methoxypyridin-4-ol 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
66080-45-3
:
Formula:
C₆H₇NO₂
MDL Number:
MFCD12546434
:
125.125 g/mol
Storage Temperature:
Ambient