0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2-Ethynyl-3-methoxypyrazine
2-Ethynyl-3-methoxypyrazine
2-Ethynyl-3-methoxypyrazine
:
Apollo Scientific
2-Ethynyl-3-methoxypyrazine
:
Apollo Scientific
:
1357945-86-8
:
:
C₇H₆N₂O
:
MFCD22199319
:
134.135 g/mol
:
GHS07
:
Refrigerator