Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Ethynylpyridine-2-carbonitrile
5-Ethynylpyridine-2-carbonitrile
5-Ethynylpyridine-2-carbonitrile
5-Ethynylpyridine-2-carbonitrile
APOSOR53008-250MG
:
Apollo Scientific
:
5-Ethynylpyridine-2-carbonitrile
APOSOR53008-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
128.131 g/mol
:
126…127 °C
:
GHS07
:
Refrigerator
:
1211584-19-8
:
C₈H₄N₂
:
MFCD18205188