Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Ethynyl-2-methoxypyridine
3-Ethynyl-2-methoxypyridine 97%
3-Ethynyl-2-methoxypyridine 97%
APOSOR52205-250MG
:
Apollo Scientific
:
3-Ethynyl-2-methoxypyridine 97%
APOSOR52205-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
133.147 g/mol
:
Ambient
:
1196145-21-7
:
C₈H₇NO
:
MFCD13189765