Order Entry
Germany
ContactUsLinkComponent
 
Irritant
2-Chloro-5-hydroxy-4-methylpyridine
Catalog # APOSOR51783-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Chloro-5-hydroxy-4-methylpyridine
Catalog # APOSOR51783-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :143.571 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1227502-89-7
Formula:C₆H₆ClNO
MDL Number:MFCD16610471