Order Entry
Germany
ContactUsLinkComponent
(2S,2'S,3S,3'S)-WingPhos ≥98%
(2S,2'S,3S,3'S)-WingPhos ≥98%
Catalog # APOSOR510259-100MG
Supplier:  Apollo Scientific
CAS Number:  
(2S,2'S,3S,3'S)-WingPhos ≥98%
Catalog # APOSOR510259-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :738.831 g/mol
Storage Temperature:Ambient
CAS Number:1435940-19-4
Formula:C₅₀H₄₄O₂P₂
MDL Number:MFCD31630708