Order Entry
Germany
ContactUsLinkComponent
3-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyrazine ≥98%
3-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyrazine ≥98%
Catalog # APOSOR510175-100MG
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyrazine ≥98%
Catalog # APOSOR510175-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :233.453 g/mol
Storage Temperature:Ambient
CAS Number:1936225-21-6
Formula:C₅H₂BrClN₄