(3-Dibenzothiophen-4-ylphenyl)boronic acid 99%
Catalog # APOSOR51014-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:304.171 g/mol
Storage Temperature:Ambient
CAS Number:1307859-67-1
Formula:C₁₈H₁₃BO₂S
MDL Number:MFCD22200801