Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(Benzyloxy)-2-iodobenzene
1-(Benzyloxy)-2-iodobenzene ≥98%
1-(Benzyloxy)-2-iodobenzene ≥98%
APOSOR510118-100MG
:
Apollo Scientific
:
1-(Benzyloxy)-2-iodobenzene ≥98%
APOSOR510118-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
310.13 g/mol
:
Refrigerator
:
142523-69-1
:
C₁₃H₁₁IO
:
MFCD14648490