Order Entry
Germany
ContactUsLinkComponent
tert-Butyl 6-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate ≥97%
tert-Butyl 6-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate ≥97%
Catalog # APOSOR510116-250MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 6-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate ≥97%
Catalog # APOSOR510116-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :297.148 g/mol
Storage Temperature:Refrigerator
CAS Number:1222809-40-6
Formula:C₁₂H₁₃BrN₂O₂