Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 6-iodopyrazine-2-carboxylate
Methyl 6-iodopyrazine-2-carboxylate ≥97%
Methyl 6-iodopyrazine-2-carboxylate ≥97%
APOSOR510076-250MG
:
Apollo Scientific
:
Methyl 6-iodopyrazine-2-carboxylate ≥97%
APOSOR510076-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
264.021 g/mol
:
Refrigerator
:
1603172-68-4
:
C₆H₅IN₂O₂
:
MFCD31618010