Order Entry
Germany
ContactUsLinkComponent
Ethyl 5-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl 5-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Catalog # APOSOR49062-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Catalog # APOSOR49062-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.186 g/mol
Storage Temperature:Ambient
CAS Number:1026456-01-8
Formula:C₉H₉N₃O₃
MDL Number:MFCD21607426