Order Entry
Germany
ContactUsLinkComponent
(2S)-2-Bromo-3-phenylpropanoic acid 96%
(2S)-2-Bromo-3-phenylpropanoic acid 96%
Catalog # APOSOR480663-1G
Supplier:  Apollo Scientific
CAS Number:  
(2S)-2-Bromo-3-phenylpropanoic acid 96%
Catalog # APOSOR480663-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.071 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:35016-63-8
Formula:C₉H₉BrO₂
MDL Number:MFCD00210121