tert-Butyl 4-(1-cyanocyclopropyl)-1H-indole-1-carboxylate
Catalog # APOSOR480133-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:282.337 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₁₈N₂O₂
MDL Number:MFCD30496572