tert-Butyl 3-(1-aminocyclopropyl)-1H-indole-1-carboxylate
Catalog # APOSOR480125-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:272.342 g/mol
Storage Temperature:Ambient
CAS Number:2007908-87-2
Formula:C₁₆H₂₀N₂O₂
MDL Number:MFCD29767794