Order Entry
Germany
ContactUsLinkComponent
1-(4-Bromopyridin-3-yl)cyclopropan-1-amine hydrochloride
1-(4-Bromopyridin-3-yl)cyclopropan-1-amine hydrochloride
Catalog # APOSOR480116-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Bromopyridin-3-yl)cyclopropan-1-amine hydrochloride
Catalog # APOSOR480116-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :249.535 g/mol
Storage Temperature:Ambient
Formula:C₈H₉BrN₂·ClH
MDL Number:MFCD30496525