1-(2-Bromopyridin-4-yl)cyclopropan-1-amine hydrochloride
Catalog # APOSOR480115-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:249.535 g/mol
Storage Temperature:Ambient
Formula:C₈H₉BrN₂·ClH
MDL Number:MFCD30496522