Ethyl 5-amino-1-pyridazin-3-yl-pyrazole-3-carboxylate
Catalog # APOSOR480072-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.227 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₁N₅O₂
MDL Number:MFCD29767770