Order Entry
Germany
ContactUsLinkComponent
Ethyl 5-amino-4-bromo-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Catalog # APOSOR480026-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-amino-4-bromo-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Catalog # APOSOR480026-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :304.184 g/mol
Storage Temperature:Ambient
CAS Number:1427022-04-5
Formula:C₁₁H₁₈BrN₃O₂
MDL Number:MFCD23135192