Ethyl 5-amino-4-iodo-1-sec-butyl-pyrazole-3-carboxylate
Catalog # APOSOR480016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:337.157 g/mol
Storage Temperature:Ambient
CAS Number:1427021-96-2
Formula:C₁₀H₁₆IN₃O₂
MDL Number:MFCD23135182