Ethyl 5-amino-4-bromo-1-sec-butyl-pyrazole-3-carboxylate
Catalog # APOSOR480015-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:290.157 g/mol
Storage Temperature:Ambient
CAS Number:1427013-82-8
Formula:C₁₀H₁₆BrN₃O₂
MDL Number:MFCD23135181