Order Entry
Germany
ContactUsLinkComponent
Ethyl 5-amino-4-bromo-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Catalog # APOSOR480012-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-amino-4-bromo-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Catalog # APOSOR480012-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :306.156 g/mol
Storage Temperature:Ambient
CAS Number:1427013-70-4
Formula:C₁₀H₁₆BrN₃O₃
MDL Number:MFCD23135178