Ethyl 5-amino-4-bromo-1-ethyl-pyrazole-3-carboxylate
Catalog # APOSOR480001-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:262.104 g/mol
Storage Temperature:Ambient
CAS Number:1427012-70-1
Formula:C₈H₁₂BrN₃O₂
MDL Number:MFCD23135134