0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Methyl 6-bromo-5-chloronicotinate
Methyl 6-bromo-5-chloronicotinate 97%
Methyl 6-bromo-5-chloronicotinate 97%
Irritant
Methyl 6-bromo-5-chloronicotinate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 6-bromo-5-chloronicotinate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
78686-80-3
:
Formula:
C₇H₅BrClNO₂
MDL Number:
MFCD14698098
:
250.477 g/mol
:
GHS07
Storage Temperature:
Ambient