Order Entry
Germany
ContactUsLinkComponent
Ethyl 2-[1-(prop-2-ynylamino)ethyl]benzoate hydrochloride
Ethyl 2-[1-(prop-2-ynylamino)ethyl]benzoate hydrochloride
Catalog # APOSOR475109-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-[1-(prop-2-ynylamino)ethyl]benzoate hydrochloride
Catalog # APOSOR475109-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.751 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₇NO₂·ClH
MDL Number:MFCD31695646