Ethyl 4-[[1-(4-bromophenyl)ethylamino]methyl]benzoate hydrochloride
Catalog # APOSOR475097-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:398.722 g/mol
Storage Temperature:Ambient
Formula:C₁₈H₂₀BrNO₂·ClH
MDL Number:MFCD31695752