N,3-Diphenylindan-1-amine hydrochloride
Catalog # APOSOR475082-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:321.843 g/mol
Storage Temperature:Ambient
Formula:C₂₁H₁₉N·ClH
MDL Number:MFCD31695708