Order Entry
Germany
ContactUsLinkComponent
N-[(1S)-1-(3-Bromophenyl)ethyl]aniline hydrochloride
N-[(1S)-1-(3-Bromophenyl)ethyl]aniline hydrochloride
Catalog # APOSOR475010-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[(1S)-1-(3-Bromophenyl)ethyl]aniline hydrochloride
Catalog # APOSOR475010-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :312.633 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₄BrN·ClH
MDL Number:MFCD31695647