Ethyl 2-[[(1S)-1-phenylethyl]amino]benzoate hydrochloride
Catalog # APOSOR475002-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:305.799 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₁₉NO₂·ClH
MDL Number:MFCD31695621