Order Entry
Germany
ContactUsLinkComponent
Ethyl 3-[[(1S)-1-phenylethyl]amino]benzoate hydrochloride
Ethyl 3-[[(1S)-1-phenylethyl]amino]benzoate hydrochloride
Catalog # APOSOR475001-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-[[(1S)-1-phenylethyl]amino]benzoate hydrochloride
Catalog # APOSOR475001-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :305.799 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₁₉NO₂·ClH
MDL Number:MFCD31695617