Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl 2-aminothiophene-3-carboxylate
tert-Butyl 2-aminothiophene-3-carboxylate 98%
tert-Butyl 2-aminothiophene-3-carboxylate 98%
APOSOR471669-5G
:
Apollo Scientific
:
tert-Butyl 2-aminothiophene-3-carboxylate 98%
APOSOR471669-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
199.27 g/mol
:
Ambient
:
59739-05-8
:
C₉H₁₃NO₂S
:
MFCD03095194