Order Entry
Germany
ContactUsLinkComponent
5-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)furan-2-carbaldehyde
5-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)furan-2-carbaldehyde
Catalog # APOSOR471522-5G
Supplier:  Apollo Scientific
CAS Number:  
5-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)furan-2-carbaldehyde
Catalog # APOSOR471522-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :251.001 g/mol
Storage Temperature:Ambient
CAS Number:1104637-03-7
Formula:C₁₀H₁₀BNO₆
MDL Number:MFCD12546038