Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(8H-Indeno[1,2-c]thiophen-8-yl)ethanol
2-(8H-Indeno[1,2-c]thiophen-8-yl)ethanol
2-(8H-Indeno[1,2-c]thiophen-8-yl)ethanol
2-(8H-Indeno[1,2-c]thiophen-8-yl)ethanol
Supplier:
Apollo Scientific
Certificates
Expand image
2-(8H-Indeno[1,2-c]thiophen-8-yl)ethanol
Supplier:
Apollo Scientific
About this item
CAS Number:
2006277-69-4
:
Formula:
C₁₃H₁₂OS
MDL Number:
MFCD28977371
:
216.299 g/mol
Storage Temperature:
Ambient