Order Entry
Germany
ContactUsLinkComponent
Ethyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Ethyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Catalog # APOSOR471398-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Catalog # APOSOR471398-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :292.244 g/mol
Storage Temperature:Ambient
CAS Number:2006277-72-9
Formula:C₁₃H₁₂N₂O₆
MDL Number:MFCD28977396