
4-Methyl-5-nitrobenzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:170918-28-2
:
Formula:C₈H₇N₃O₂
MDL Number:MFCD18807642
Melting Pt:259…261 °C (subl.)
:177.16 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together