Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Aminomethyl)benzoxazole hydrochloride
6-(Aminomethyl)benzoxazole hydrochloride
6-(Aminomethyl)benzoxazole hydrochloride
Irritant
6-(Aminomethyl)benzoxazole hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-(Aminomethyl)benzoxazole hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
1956341-07-3
:
Formula:
C₈H₈N₂O·ClH
MDL Number:
MFCD23703073
:
184.623 g/mol
:
GHS07
Storage Temperature:
Freezer