Diethyl 2-(4-Chloro-2-nitrophenyl)malonate
Catalog # APOSOR471130-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:315.706 g/mol
Storage Temperature:Ambient
CAS Number:10565-16-9
Formula:C₁₃H₁₄ClNO₆
MDL Number:MFCD06204600