Diethyl 2-(2-Bromo-4-pyridyl)malonate
Catalog # APOSOR471129-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:316.148 g/mol
Storage Temperature:Ambient
CAS Number:1387560-71-5
Formula:C₁₂H₁₄BrNO₄
MDL Number:MFCD28154652