Order Entry
Germany
ContactUsLinkComponent
Methyl 2-(2-Bromo-5-methyl-4-thiazolyl)acetate
Methyl 2-(2-Bromo-5-methyl-4-thiazolyl)acetate
Catalog # APOSOR471085-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-(2-Bromo-5-methyl-4-thiazolyl)acetate
Catalog # APOSOR471085-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.113 g/mol
Storage Temperature:Ambient
CAS Number:496062-15-8
Formula:C₇H₈BrNO₂S
MDL Number:MFCD09038182