Order Entry
Germany
ContactUsLinkComponent
Ethyl 1-Ethyl-2,4-dioxopiperidine-3-carboxylate
Ethyl 1-Ethyl-2,4-dioxopiperidine-3-carboxylate
Catalog # APOSOR471061-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-Ethyl-2,4-dioxopiperidine-3-carboxylate
Catalog # APOSOR471061-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :213.23 g/mol
Storage Temperature:Ambient
CAS Number:104966-05-4
Formula:C₁₀H₁₅NO₄
MDL Number:MFCD28122817