Order Entry
Germany
ContactUsLinkComponent
3-Bromo-7-chloro-1H-pyrazolo[4,3-d]pyrimidine
3-Bromo-7-chloro-1H-pyrazolo[4,3-d]pyrimidine
Catalog # APOSOR470800-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-7-chloro-1H-pyrazolo[4,3-d]pyrimidine
Catalog # APOSOR470800-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.453 g/mol
Storage Temperature:Ambient
CAS Number:68510-70-3
Formula:C₅H₂BrClN₄
MDL Number:MFCD09746269