Methyl (1R,2S,3S,4S,6R)-5-oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Catalog # APOSOR470727-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.174 g/mol
Storage Temperature:Ambient
CAS Number:1932427-01-4
Formula:C₉H₁₀O₃
MDL Number:MFCD28096495