Diethyl 2-(4-Methoxybenzyl)malonate
Catalog # APOSOR470705-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:280.316 g/mol
Storage Temperature:Ambient
CAS Number:6335-37-1
Formula:C₁₅H₂₀O₅
MDL Number:MFCD00157074